6-Fluoro-3-methoxypyridin-2-amine 97%

Lieferant: Apollo Scientific
Achtung

PC535050-250MG
APOSPC535050-250MGEA 97.3 EUR
APOSPC535050-250MG
6-Fluoro-3-methoxypyridin-2-amine 97%
6-Fluoro-3-methoxypyridin-2-amine
Formel: C₆H₇FN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD28722398
CAS-Nummer: 1805602-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR