4-Fluoro-3-methylpyridin-2-amine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC535051-250MG
APOSPC535051-250MGEA
120
EUR
APOSPC535051-250MG
4-Fluoro-3-methylpyridin-2-amine 97%
4-Fluoro-3-methylpyridin-2-amine
Formel:
C₆H₇FN₂ Molecular Weight: 126,132 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16606166 CAS-Nummer: 1227586-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...