3-Methoxy-4-(trifluoromethyl)pyridin-2-amine 97%

Lieferant: Apollo Scientific
Achtung

PC535052-250MG
APOSPC535052-250MGEA 0 EUR
APOSPC535052-250MG
3-Methoxy-4-(trifluoromethyl)pyridin-2-amine 97%
3-Methoxy-4-(trifluoromethyl)pyridin-2-amine
Formel: C₇H₇F₃N₂O
Lagertemperatur: Kühlschrank
MDL: MFCD16606204
CAS-Nummer: 1227581-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR