3-Bromo-6-(difluoromethyl)pyridin-2-amine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC535054-250MG
APOSPC535054-250MGEA
0
EUR
APOSPC535054-250MG
3-Bromo-6-(difluoromethyl)pyridin-2-amine 97%
3-Bromo-6-(difluoromethyl)pyridin-2-amine
Formel:
C₆H₅BrF₂N₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD25476828 CAS-Nummer: 1805028-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...