3-Methoxy-5-(trifluoromethyl)pyridin-2-amine 95%

Lieferant: Apollo Scientific
Achtung

PC535055-100MG
APOSPC535055-100MGEA 56.7 EUR
APOSPC535055-100MG
3-Methoxy-5-(trifluoromethyl)pyridin-2-amine 95%
3-Methoxy-5-(trifluoromethyl)pyridin-2-amine
Formel: C₇H₇F₃N₂O
Molecular Weight: 192,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606205
CAS-Nummer: 1227603-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR