5-Bromo-2-methyl-6-(trifluoromethyl)pyrimidin-4-ol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC535058-250MG
APOSPC535058-250MGEA
105
EUR
APOSPC535058-250MG
5-Bromo-2-methyl-6-(trifluoromethyl)pyrimidin-4-ol 95%
5-Bromo-2-methyl-6-(trifluoromethyl)pyrimidin-4-ol
Formel:
C₆H₄BrF₃N₂O Lagertemperatur: Kühlschrank |
MDL:
MFCD16988234 CAS-Nummer: 1240602-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...