3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC535059-100MG
PC535059-250MG
APOSPC535059-100MGEA
267
EUR
APOSPC535059-100MG
APOSPC535059-250MG
3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide 95%
3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide
Formel:
C₇H₅BrF₃N·BrH Molecular Weight: 320.932 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD29760335 CAS-Nummer: 1384972-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...