3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide 95%

Lieferant: Apollo Scientific
Achtung

PC535059-100MG PC535059-250MG
APOSPC535059-100MGEA 0 EUR
APOSPC535059-100MG APOSPC535059-250MG
3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide 95%
3-(Bromomethyl)-5-(trifluoromethyl)pyridine hydrobromide
Formel: C₇H₅BrF₃N·BrH
Lagertemperatur: Kühlschrank
MDL: MFCD29760335
CAS-Nummer: 1384972-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR