2-Bromo-5-fluoroisonicotinamide 97%

Lieferant: Apollo Scientific
Achtung

PC535060-1G PC535060-250MG PC535060-5G
APOSPC535060-1GEA 95.7 EUR
APOSPC535060-1G APOSPC535060-250MG APOSPC535060-5G
2-Bromo-5-fluoroisonicotinamide 97%
2-Bromo-5-fluoroisonicotinamide
Formel: C₆H₄BrFN₂O
Molecular Weight: 219.011 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30342404
CAS-Nummer: 1799792-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR