2-Bromo-5-fluoroisonicotinamide 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC535060-1G
PC535060-250MG
PC535060-5G
APOSPC535060-1GEA
95.7
EUR
APOSPC535060-1G
APOSPC535060-250MG
APOSPC535060-5G
2-Bromo-5-fluoroisonicotinamide 97%
2-Bromo-5-fluoroisonicotinamide
Formel:
C₆H₄BrFN₂O Molecular Weight: 219.011 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD30342404 CAS-Nummer: 1799792-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...