4-Bromo-6-fluoroisoquinolin-1(2H)-one 95+%

Lieferant: Apollo Scientific
Achtung

PC535064-250MG
APOSPC535064-250MGEA 98.6 EUR
APOSPC535064-250MG
4-Bromo-6-fluoroisoquinolin-1(2H)-one 95+%
4-Bromo-6-fluoroisoquinolin-1(2H)-one
Formel: C₉H₅BrFNO
Lagertemperatur: Kühlschrank
MDL: MFCD14706174
CAS-Nummer: 1227607-99-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR