4-Fluoro-2-methyl-1H-indole 95+%

Lieferant: Apollo Scientific
Achtung

PC535067-250MG
APOSPC535067-250MGEA 92.8 EUR
APOSPC535067-250MG
4-Fluoro-2-methyl-1H-indole 95+%
4-Fluoro-2-methyl-1H-indole
Formel: C₉H₈FN
Molecular Weight: 149,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18380834
CAS-Nummer: 1260383-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR