3-Chloro-5-(2-fluoroethoxy)pyridazine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC535069-250MG
APOSPC535069-250MGEA
87
EUR
APOSPC535069-250MG
3-Chloro-5-(2-fluoroethoxy)pyridazine 95+%
3-Chloro-5-(2-fluoroethoxy)pyridazine
Formel:
C₆H₆ClFN₂O Lagertemperatur: Kühlschrank |
MDL:
MFCD19688855 CAS-Nummer: 1346691-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...