Methyl 2-fluoro-4-(methylsulphonyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC53508-1G PC53508-250MG PC53508-5G
APOSPC53508-1GEA 0 EUR
APOSPC53508-1G APOSPC53508-250MG APOSPC53508-5G
Methyl 2-fluoro-4-(methylsulphonyl)benzoate
Methyl 2-fluoro-4-(methylsulphonyl)benzoate
Formel: C₉H₉FO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD27578943
CAS-Nummer: 1820716-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR