7-Bromo-5-fluoro-3,4-dihydroisoquinolin-1(2H)-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC535081-250MG
APOSPC535081-250MGEA
108
EUR
APOSPC535081-250MG
7-Bromo-5-fluoro-3,4-dihydroisoquinolin-1(2H)-one 95%
7-Bromo-5-fluoro-3,4-dihydroisoquinolin-1(2H)-one
Formel:
C₉H₇BrFNO Molecular Weight: 244,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20287127 CAS-Nummer: 1341674-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...