5-Fluoro-1,2,3,4-tetrahydronaphthalen-1-amine 95+%

Lieferant: Apollo Scientific
Achtung

PC535087-250MG
APOSPC535087-250MGEA 133 EUR
APOSPC535087-250MG
5-Fluoro-1,2,3,4-tetrahydronaphthalen-1-amine 95+%
5-Fluoro-1,2,3,4-tetrahydronaphthalen-1-amine
Formel: C₁₀H₁₂FN
Molecular Weight: 165,207 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07373975
CAS-Nummer: 907973-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR