5-Bromo-7-fluoroisobenzofuran-1(3H)-one 95%

Lieferant: Apollo Scientific
Achtung

PC535089-250MG
APOSPC535089-250MGEA 288 EUR
APOSPC535089-250MG
5-Bromo-7-fluoroisobenzofuran-1(3H)-one 95%
5-Bromo-7-fluoroisobenzofuran-1(3H)-one
Formel: C₈H₄BrFO₂
Molecular Weight: 231,019 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28962614
CAS-Nummer: 1255208-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR