4-Chloro-7-fluoro-2,3-dihydro-1H-inden-1-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC535090-250MG
APOSPC535090-250MGEA
62.5
EUR
APOSPC535090-250MG
4-Chloro-7-fluoro-2,3-dihydro-1H-inden-1-one 97%
4-Chloro-7-fluoro-2,3-dihydro-1H-inden-1-one
Formel:
C₉H₆ClFO Molecular Weight: 184,595 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18208182 CAS-Nummer: 1260018-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...