2-Bromo-6-fluorobenzal fluoride

Lieferant: Apollo Scientific
Achtung

PC53521-1G PC53521-5G
APOSPC53521-1GEA 75.4 EUR
APOSPC53521-1G APOSPC53521-5G
2-Bromo-6-fluorobenzal fluoride
2-Bromo-6-fluorobenzal fluoride
Formel: C₇H₄BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD14698533
CAS-Nummer: 1166820-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR