4-Amino-3-(4-fluorophenyl)butyric acid methyl ester hydrochloride

Lieferant: Apollo Scientific
Achtung

PC53523-1G PC53523-250MG PC53523-50MG
APOSPC53523-1GEA 720 EUR
APOSPC53523-1G APOSPC53523-250MG APOSPC53523-50MG
4-Amino-3-(4-fluorophenyl)butyric acid methyl ester hydrochloride
4-Amino-3-(4-fluorophenyl)butyric acid methyl ester hydrochloride
Formel: C₁₁H₁₄FNO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD30492328
CAS-Nummer: 2149591-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR