Potassium 2-fluorobenzyl-trifluoroborate

Lieferant: Apollo Scientific
Achtung

PC53537-1G PC53537-5G PC53537-250MG
APOSPC53537-1GEA 0 EUR
APOSPC53537-1G APOSPC53537-5G APOSPC53537-250MG
Potassium 2-fluorobenzyl-trifluoroborate
Potassium 2-fluorobenzyl-trifluoroborate
Formel: C₇H₆BF₄·K
Lagertemperatur: Raumtemperatur
MDL: MFCD09993122
CAS-Nummer: 1246073-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR