6-Chloro-2-(trifluoromethyl)pyrimidin-4-amine 98%

Lieferant: Apollo Scientific
Achtung

PC53540-250MG PC53540-1G
APOSPC53540-250MGEA 0 EUR
APOSPC53540-250MG APOSPC53540-1G
6-Chloro-2-(trifluoromethyl)pyrimidin-4-amine 98%
6-Chloro-2-(trifluoromethyl)pyrimidin-4-amine
Formel: C₅H₃ClF₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08436599
CAS-Nummer: 1480-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR