6-Chloro-2-(trifluoromethyl)pyrimidin-4-amine 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC53540-250MG
PC53540-1G
APOSPC53540-250MGEA
0
EUR
APOSPC53540-250MG
APOSPC53540-1G
6-Chloro-2-(trifluoromethyl)pyrimidin-4-amine 98%
6-Chloro-2-(trifluoromethyl)pyrimidin-4-amine
Formel:
C₅H₃ClF₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436599 CAS-Nummer: 1480-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...