2-Brom-6-chlor-4-(trifluormethoxy)anilin

Lieferant: Apollo Scientific
Achtung

PC53547-1G PC53547-5G PC53547-25G
APOSPC53547-1GEA 0 EUR
APOSPC53547-1G APOSPC53547-5G APOSPC53547-25G
2-Brom-6-chlor-4-(trifluormethoxy)anilin
2-Brom-6-chlor-4-(trifluormethoxy)anilin
Formel: C₇H₄BrClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD16618925
CAS-Nummer: 1228108-74-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR