2,4-Difluoro-5-methylbenzenesulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC53548-1G
PC53548-250MG
APOSPC53548-1GEA
400
EUR
APOSPC53548-1G
APOSPC53548-250MG
2,4-Difluoro-5-methylbenzenesulphonyl chloride 97%
2,4-Difluoro-5-methylbenzenesulphonyl chloride
Formel:
C₇H₅ClF₂O₂S Molecular Weight: 226.628 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22196689 CAS-Nummer: 1235407-51-8 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...