Pentafluoropropionaldehyde hydrate

Lieferant: Apollo Scientific
Achtung

PC5355-25G PC5355-5G
APOSPC5355-25GEA 577 EUR
APOSPC5355-25G APOSPC5355-5G
Pentafluoropropionaldehyde hydrate
Pentafluoropropionaldehyde hydrate
Formel: C₃H₃F₅O₂·xH₂O
Siedepunkt: 93…94 °C
Schmelzpunkt: 53…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00792902
CAS-Nummer: 422-63-9
EINECS: 207-020-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR