2,4-Difluoro-DL-phenylglycine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-DL-2,4-DiF-Phg-OH, 2-Amino-2-(2,4-difluorophenyl)ethanoic acid, 2-Amino-2-(2,4-difluorophenyl)acetic acid, Amino(2,4-difluorophenyl)acetic acid

PC5357-1G PC5357-250MG PC5357-5G
APOSPC5357-1GEA 50.9 EUR
APOSPC5357-1G APOSPC5357-250MG APOSPC5357-5G
2,4-Difluoro-DL-phenylglycine 98%
2,4-Difluoro-DL-phenylglycine
Formel: C₈H₇F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02662503
CAS-Nummer: 240409-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR