3-Chloro-1-hydroxy-5-(trifluoromethyl)pyridin-2(1H)-one

Lieferant: Apollo Scientific
Achtung

PC53581-1G
APOSPC53581-1GEA 241 EUR
APOSPC53581-1G
3-Chloro-1-hydroxy-5-(trifluoromethyl)pyridin-2(1H)-one
3-Chloro-1-hydroxy-5-(trifluoromethyl)pyridin-2(1H)-one
Formel: C₆H₃ClF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29042535
CAS-Nummer: 1823183-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR