4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Mercapto-6-(trifluoromethyl)-4-pyrimidinol, 4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol, 4-Hydroxy-2-mercapto-6-(trifluoromethyl)pyrimidine
PC53582-1G
APOSPC53582-1GEA
21.9
EUR
APOSPC53582-1G
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol 95+%
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol
Formel:
C₅H₃F₃N₂OS Molecular Weight: 196,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00112978 CAS-Nummer: 368-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...