Methyl 4-chloro-2-fluoro-5-nitro-benzoate 98%

Lieferant: Apollo Scientific

PC53609-1G PC53609-250MG PC53609-5G
APOSPC53609-1GEA 0 EUR
APOSPC53609-1G APOSPC53609-250MG APOSPC53609-5G
Methyl 4-chloro-2-fluoro-5-nitro-benzoate 98%
Methyl 4-chloro-2-fluoro-5-nitro-benzoate
Formel: C₈H₅ClFNO₄
Lagertemperatur: Kühlschrank
MDL: MFCD12172989
CAS-Nummer: 1070893-15-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR