3-Bromo-2-chloro-6-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC5361-1G
PC5361-250MG
PC5361-5G
APOSPC5361-1GEA
128
EUR
APOSPC5361-1G
APOSPC5361-250MG
APOSPC5361-5G
3-Bromo-2-chloro-6-(trifluoromethyl)pyridine
3-Bromo-2-chloro-6-(trifluoromethyl)pyridine
Formel:
C₆H₂BrClF₃N Schmelzpunkt: 39…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226615 CAS-Nummer: 1159512-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...