Methyl 4,5-difluoro-2-nitro-benzoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 4,5-difluoro-2-nitrobenzoate

PC53622-25G
APOSPC53622-25GEA 78.3 EUR
APOSPC53622-25G
Methyl 4,5-difluoro-2-nitro-benzoate 95%
Methyl 4,5-difluoro-2-nitro-benzoate
Formel: C₈H₅F₂NO₄
Molecular Weight: 217,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11042825
CAS-Nummer: 1015433-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR