3-(Difluormethoxy)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(Difluoromethoxy)phenyl)methanol

PC5371-1G PC5371-5G
APOSPC5371-1GEA 185 EUR
APOSPC5371-1G APOSPC5371-5G
3-(Difluormethoxy)benzylalkohol 98%
3-(Difluormethoxy)benzylalkohol
Formel: C₈H₈F₂O₂
Siedepunkt: 95…100 °C/3,3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00236227
CAS-Nummer: 125903-81-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR