2-(Difluoromethyl)pyridin-4-amine > 97%
Lieferant: Apollo Scientific
PC540000-250MG
APOSPC540000-250MGEA
304
EUR
APOSPC540000-250MG
2-(Difluoromethyl)pyridin-4-amine > 97%
2-(Difluoromethyl)pyridin-4-amine
Formel:
C₆H₆F₂N₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD25476532 CAS-Nummer: 1446509-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...