3-Chloro-5-fluoropyridin-2(1H)-one ≥97%
Lieferant: Apollo Scientific
PC540001-250MG
APOSPC540001-250MGEA
31.9
EUR
APOSPC540001-250MG
3-Chloro-5-fluoropyridin-2(1H)-one ≥97%
3-Chloro-5-fluoropyridin-2(1H)-one
Formel:
C₅H₃ClFNO Lagertemperatur: Kühlschrank |
MDL:
MFCD13185439 CAS-Nummer: 1214345-43-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...