4-Amino-5-fluoro-2-methylbenzonitrile ≥97%
Lieferant: Apollo Scientific
PC540002-250MG
APOSPC540002-250MGEA
186
EUR
APOSPC540002-250MG
4-Amino-5-fluoro-2-methylbenzonitrile ≥97%
4-Amino-5-fluoro-2-methylbenzonitrile
Formel:
C₈H₇FN₂ Molecular Weight: 150,153 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD30340362 CAS-Nummer: 1849279-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...