6-Amino-5-fluoroindolin-2-one ≥97%

Lieferant: Apollo Scientific

PC540003-250MG
APOSPC540003-250MGEA 87 EUR
APOSPC540003-250MG
6-Amino-5-fluoroindolin-2-one ≥97%
6-Amino-5-fluoroindolin-2-one
Formel: C₈H₇FN₂O
Molecular Weight: 166,152 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12924695
CAS-Nummer: 150544-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR