(S)-1-(5-Fluoropyrimidin-2-yl)ethanamine hydrochloride 98%
Lieferant: Apollo Scientific
PC540007-250MG
APOSPC540007-250MGEA
188
EUR
APOSPC540007-250MG
(S)-1-(5-Fluoropyrimidin-2-yl)ethanamine hydrochloride 98%
(S)-1-(5-Fluoropyrimidin-2-yl)ethanamine hydrochloride
Formel:
C₆H₈FN₃·ClH Molecular Weight: 177,607 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18207119 CAS-Nummer: 935667-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...