4-Fluoro-1H-indole-5-carboxylic acid ≥98%

Lieferant: Apollo Scientific

PC540008-250MG
APOSPC540008-250MGEA 318 EUR
APOSPC540008-250MG
4-Fluoro-1H-indole-5-carboxylic acid ≥98%
4-Fluoro-1H-indole-5-carboxylic acid
Formel: C₉H₆FNO₂
Molecular Weight: 179,148 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12924428
CAS-Nummer: 908600-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR