6-Fluorobenzofuran-3-carboxylic acid ≥95%

Lieferant: Apollo Scientific

PC540009-250MG
APOSPC540009-250MGEA 114 EUR
APOSPC540009-250MG
6-Fluorobenzofuran-3-carboxylic acid ≥95%
6-Fluorobenzofuran-3-carboxylic acid
Formel: C₉H₅FO₃
Molecular Weight: 180,133 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22544066
CAS-Nummer: 1393561-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR