(6,7-Difluoro-1H-benzo[d]imidazol-2-yl)methanamine ≥98%

Lieferant: Apollo Scientific

PC540010-250MG
APOSPC540010-250MGEA 302 EUR
APOSPC540010-250MG
(6,7-Difluoro-1H-benzo[d]imidazol-2-yl)methanamine ≥98%
(6,7-Difluoro-1H-benzo[d]imidazol-2-yl)methanamine
Formel: C₈H₇F₂N₃
Molecular Weight: 183,158 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD22564762
CAS-Nummer: 1201769-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR