Methyl 4-amino-2-fluoro-3-methylbenzoate ≥97%

Lieferant: Apollo Scientific

PC540011-250MG
APOSPC540011-250MGEA 115 EUR
APOSPC540011-250MG
Methyl 4-amino-2-fluoro-3-methylbenzoate ≥97%
Methyl 4-amino-2-fluoro-3-methylbenzoate
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13184288
CAS-Nummer: 1206680-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR