3,4-Difluoro-2-formylphenylboronic acid ≥98%
Lieferant: Apollo Scientific
PC540012-250MG
APOSPC540012-250MGEA
128
EUR
APOSPC540012-250MG
3,4-Difluoro-2-formylphenylboronic acid ≥98%
3,4-Difluoro-2-formylphenylboronic acid
Formel:
C₇H₅BF₂O₃ Molecular Weight: 185,921 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD23713092 CAS-Nummer: 1451392-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...