5-Bromo-6-fluoropyridin-2(1H)-one ≥95%
Lieferant: Apollo Scientific
PC540013-250MG
APOSPC540013-250MGEA
84.1
EUR
APOSPC540013-250MG
5-Bromo-6-fluoropyridin-2(1H)-one ≥95%
5-Bromo-6-fluoropyridin-2(1H)-one
Formel:
C₅H₃BrFNO Lagertemperatur: Kühlschrank |
MDL:
MFCD16609912 CAS-Nummer: 1227597-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...