2-Bromo-5-fluoroisonicotinonitrile ≥97%
Lieferant: Apollo Scientific
PC540016-250MG
APOSPC540016-250MGEA
278
EUR
APOSPC540016-250MG
2-Bromo-5-fluoroisonicotinonitrile ≥97%
2-Bromo-5-fluoroisonicotinonitrile
Formel:
C₆H₂BrFN₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD18258644 CAS-Nummer: 1256822-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...