Methyl 5-chloro-2-fluoro-4-hydroxybenzoate ≥98%
Lieferant: Apollo Scientific
PC540017-250MG
APOSPC540017-250MGEA
63.8
EUR
APOSPC540017-250MG
Methyl 5-chloro-2-fluoro-4-hydroxybenzoate ≥98%
Methyl 5-chloro-2-fluoro-4-hydroxybenzoate
Formel:
C₈H₆ClFO₃ Molecular Weight: 204,583 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22371002 CAS-Nummer: 245743-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...