2-Chloro-4-(2,2,2-trifluoroethoxy)pyrimidine > 97%

Lieferant: Apollo Scientific

PC540018-250MG
APOSPC540018-250MGEA 94.4 EUR
APOSPC540018-250MG
2-Chloro-4-(2,2,2-trifluoroethoxy)pyrimidine > 97%
2-Chloro-4-(2,2,2-trifluoroethoxy)pyrimidine
Formel: C₆H₄ClF₃N₂O
Molecular Weight: 212,557 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD16712374
CAS-Nummer: 935252-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR