5-(Trifluoromethyl)-1H-indole-3-carbaldehyde ≥98%

Lieferant: Apollo Scientific

PC540019-250MG
APOSPC540019-250MGEA 125 EUR
APOSPC540019-250MG
5-(Trifluoromethyl)-1H-indole-3-carbaldehyde ≥98%
5-(Trifluoromethyl)-1H-indole-3-carbaldehyde
Formel: C₁₀H₆F₃NO
Molecular Weight: 213,156 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09954785
CAS-Nummer: 468718-16-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR