Methyl 3-fluoro-2-(trifluoromethyl)isonicotinate > 98%

Lieferant: Apollo Scientific

PC540020-250MG
APOSPC540020-250MGEA 377 EUR
APOSPC540020-250MG
Methyl 3-fluoro-2-(trifluoromethyl)isonicotinate > 98%
Methyl 3-fluoro-2-(trifluoromethyl)isonicotinate
Formel: C₈H₅F₄NO₂
Molecular Weight: 223,124 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16611619
CAS-Nummer: 1227594-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR