6-Bromo-4-fluoroquinoline ≥97%

Lieferant: Apollo Scientific

PC540021-250MG
APOSPC540021-250MGEA 195 EUR
APOSPC540021-250MG
6-Bromo-4-fluoroquinoline ≥97%
6-Bromo-4-fluoroquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226,045 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29921328
CAS-Nummer: 1713240-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR